Methyl 5-cyano-2-methoxynicotinate 97%

Supplier: Ambeed
Warning

A1003262-100MG A1003262-250MG A1003262-50MG A1003262-1G A1003262-5G
103795-370EA 91 USD
103795-370 103821-072 76646-072 76646-074 76646-076 77296-640 77296-642
Methyl 5-cyano-2-methoxynicotinate 97%
Methyl 5-cyano-2-methoxynicotinate

Precaución: for Research Use Only

Formula: C₉H₈N₂O₃
Boiling Pt: 311.0±42.0 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD22371018
CAS Number: 1806320-48-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR