Methyl-5-chloropicolinate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 5-chloropyridine-2-carboxylate
A231690-100G
A231690-10G
A231690-250MG
A231690-1G
103810-786EA
9
USD
103810-786
103850-540
76771-172
76771-174
76771-176
77089-432
Methyl-5-chloropicolinate 98%
Methyl 5-chloropicolinate
Precaución: for Research Use Only
Formula:
C₇H₆ClNO₂ MW: 171.5800 g/mol Flash Pt: 103.4 °C Storage Temperature: Ambient |
MDL Number:
MFCD09702466 CAS Number: 132308-19-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...