Methyl-5-bromopyrimidine-4-carboxylate 98%
Supplier: Ambeed
|
Warning
|
A303590-100MG
A303590-250MG
A303590-5G
103814-382EA
77
USD
103814-382
103835-548
76787-800
76787-802
77305-874
Methyl-5-bromopyrimidine-4-carboxylate 98%
Methyl-5-bromopyrimidine-4-carboxylate
Precaución: for Research Use Only
Formula:
C₆H₅BrN₂O₂ Boiling Pt: 279.7 °C (760 mm Hg) Flash Pt: 123 °C Storage Temperature: Ambient |
MDL Number:
MFCD11111841 CAS Number: 1009826-93-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...