Methyl-5-bromopyrimidine-2-carboxyate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 5-Bromopyrimidine-2-carboxylate, 5-Bromopyrimidine-2-carboxylic acid methyll ester

A281111-100MG A281111-10G A281111-250MG A281111-1G A281111-25G A281111-5G
103813-536EA 14 USD
103813-536 103853-488 76787-396 76787-398 76787-400 77305-378 77305-380 77305-382
Methyl-5-bromopyrimidine-2-carboxyate 95%
Methyl-5-bromopyrimidine-2-carboxyate

Precaución: for Research Use Only

Formula: C₆H₅BrN₂O₂
MW: 217.02 g/mol
Boiling Pt: 314.4 °C (760 mm Hg)
Flash Pt: 143.9 °C
Storage Temperature: Ambient
MDL Number: MFCD11111606
CAS Number: 89581-38-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR