Methyl-5-bromopicolinate 98%

Supplier: Ambeed
Warning

Synonyms: 5-Bromopyridine-2-carboxylic acid methyl ester, Methyl 5-bromopyridine-2-carboxylate, Methyl 5-bromopicolinate

A155749-100G A155749-10G A155749-500G A155749-1G A155749-25G A155749-5G
103804-794EA 9 USD
103804-794 103842-996 76997-524 76997-526 76997-528 77318-726 77318-728 77318-730
Methyl-5-bromopicolinate 98%
Methyl 5-bromopicolinate

Precaución: for Research Use Only

Formula: C₇H₆BrNO₂
Boiling Pt: 290.9 °C (760 mm Hg)
Melting Pt: 101…103 °C
Flash Pt: 129.7 °C
Storage Temperature: Ambient
MDL Number: MFCD04112493
CAS Number: 29682-15-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR