Methyl-5-bromonicotinate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 5-bromonicotinate
A181461-100G
A181461-10G
A181461-1G
A181461-500G
A181461-25G
A181461-5G
103845-982EA
6
USD
103845-982
76769-550
76769-552
76769-554
76769-556
77083-506
77301-720
Methyl-5-bromonicotinate 98%
Methyl-5-bromonicotinate
Precaución: for Research Use Only
Formula:
C₇H₆BrNO₂ Boiling Pt: 241.7 °C (760 mm Hg) Melting Pt: 96…100 °C Flash Pt: 100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00173668 CAS Number: 29681-44-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...