Methyl-5-bromo-2-furoate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Bromo-2-furoic acid methyll ester, 5-Bromo-2-furancarboxylic acid methyll ester, Methyl 5-Bromo-2-furancarboxylate, Methyl 5-Bromo-2-furoate, Methyl 5-bromofuran-2-carboxylate
A122402-100G
A122402-10G
A122402-25G
A122402-1G
A122402-5G
103800-226EA
5
USD
103800-226
77094-540
77094-542
77094-544
77312-356
77312-358
Methyl-5-bromo-2-furoate 97%
Methyl-5-bromo-2-furoate
Precaución: for Research Use Only
Formula:
C₆H₅BrO₃ MW: 205.01 g/mol Boiling Pt: 226.3 °C (760 mm Hg) Melting Pt: 63…68 °C Flash Pt: 90.7 °C Storage Temperature: Ambient |
MDL Number:
MFCD00092313 CAS Number: 2527-99-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...