Methyl 5-bromobenzo[d]oxazole-2-carboxylate 97%
Supplier: Ambeed
|
Warning
|
A157930-100MG
A157930-250MG
103805-064EA
429
USD
103805-064
103843-270
76973-394
77671-238
Methyl 5-bromobenzo[d]oxazole-2-carboxylate 97%
Methyl 5-bromobenzo[d]oxazole-2-carboxylate
Precaución: for Research Use Only
Formula:
C₉H₆BrNO₃ MW: 256.05 g/mol Boiling Pt: 330.4±34.0 °C (760 mm Hg) Flash Pt: 153.6 °C Storage Temperature: Ambient |
MDL Number:
MFCD09832200 CAS Number: 954239-61-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...