Methyl 5-bromo-6-oxo-1,6-dihydropyridine-2-carboxylate 98%

Supplier: Ambeed
Warning

A206757-100MG A206757-250MG A206757-1G A206757-5G
103817-728EA 44 USD
103817-728 103848-404 76825-760 76825-762 77303-002 77303-004
Methyl 5-bromo-6-oxo-1,6-dihydropyridine-2-carboxylate 98%
Methyl 5-bromo-6-oxo-1,6-dihydropyridine-2-carboxylate

Precaución: for Research Use Only

Formula: C₇H₆BrNO₃
MW: 232.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16036911
CAS Number: 178876-86-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR