Methyl 5-bromo-6-chloropicolinate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 5-bromo-6-chloropyridine-2-carboxylate
A252061-250MG
A252061-1G
A252061-5G
103812-020EA
103
USD
103812-020
103851-862
76826-978
77091-262
77304-570
Methyl 5-bromo-6-chloropicolinate 97%
Methyl 5-bromo-6-chloropicolinate
Precaución: for Research Use Only
Formula:
C₇H₅BrClNO₂ MW: 250.48 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD14698104 CAS Number: 1214353-79-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...