Methyl-5-bromo-6-chloronicotinate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 5-bromo-6-chloronicotinate
A133326-100G
A133326-10G
A133326-1G
A133326-25G
A133326-5G
103801-860EA
9
USD
103801-860
76870-854
76870-856
77298-622
77314-500
77314-502
Methyl-5-bromo-6-chloronicotinate 98%
Methyl-5-bromo-6-chloronicotinate
Precaución: for Research Use Only
Formula:
C₇H₅BrClNO₂ MW: 250.48 g/mol Melting Pt: 76…79 °C Flash Pt: 122.7 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03844855 CAS Number: 78686-77-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...