Methyl-5-bromo-4-methylpicolinate 96%

Supplier: Ambeed
Warning

Synonyms: Methyl 5-bromo-4-methylpicolinate, 5-Bromo-4-methyl-pyridine-2-carboxylic acid methyl ester

A126868-100MG A126868-10G A126868-250MG A126868-1G A126868-25G A126868-5G
103800-866EA 92 USD
103800-866 77009-698 77009-700 77009-702 77313-100 77313-102 77313-104
Methyl-5-bromo-4-methylpicolinate 96%
Methyl-5-bromo-4-methylpicolinate

Precaución: for Research Use Only

Formula: C₈H₈BrNO₂
MW: 230.0600 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07375102
CAS Number: 886365-06-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR