Methyl 5-bromo-4-methoxypyridine-2-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 5-bromo-4-methoxypicolinate

A272911-100MG A272911-250MG A272911-1G A272911-25G
103813-114EA 12 USD
103813-114 103853-028 77014-446 77014-448 77092-814 77092-816
Methyl 5-bromo-4-methoxypyridine-2-carboxylate 97%
Methyl 5-bromo-4-methoxypyridine-2-carboxylate

Precaución: for Research Use Only

Formula: C₈H₈BrNO₃
MW: 246.058 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18257513
CAS Number: 1256789-95-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR