Methyl-5-bromo-3-methylpicolinate 98%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-3-methylpyridine-2-carboxylic acid methyl ester, Methyl 5-bromo-3-methylpyridine-2-carboxylate, Methyl 5-bromo-3-methylpicolinate

A393639-1G A393639-25G A393639-5G A393639-10G A393639-250MG
103826-928EA 16 USD
103826-928 103837-966 76841-370 76841-372 76841-374 77016-228 77016-230
Methyl-5-bromo-3-methylpicolinate 98%
Methyl 5-bromo-3-methylpicolinate

Precaución: for Research Use Only

Formula: C₈H₈BrNO₃
MW: 230.0586 g/mol
Boiling Pt: 302.2 °C (760 mm Hg)
Flash Pt: 136.6 °C
Storage Temperature: Ambient
MDL Number: MFCD07375126
CAS Number: 213771-32-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR