Methyl 5-bromo-3-chloropyridine-2-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-3-chloro-2-(methoxycarbonyl)pyridine, Methyl 5-bromo-3-chloropicolinate

A235937-250MG A235937-1G A235937-25G A235937-5G
103811-048EA 11 USD
103811-048 103850-818 76987-504 77089-840 77089-842 77089-844
Methyl 5-bromo-3-chloropyridine-2-carboxylate 97%
Methyl 5-bromo-3-chloropyridine-2-carboxylate

Precaución: for Research Use Only

Formula: C₇H₅BrClNO₂
MW: 250.477 g/mol
Boiling Pt: 311 °C (760 mm Hg)
Flash Pt: 142 °C
Storage Temperature: Ambient
MDL Number: MFCD14698107
CAS Number: 1214336-41-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR