Methyl-5-bromo-2-hydroxynicotinate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 5-bromo-2-oxo-1,2-dihydropyridine-3-carboxylate, methyl 5-bromo-2-hydroxypyridine-3-carboxylate, Methyl 5-bromo-2-oxo-1,2-dihydro-3-pyridinecarboxylate

A256330-100G A256330-10G A256330-250MG A256330-25G A256330-5G A256330-1G
103812-262EA 8 USD
103812-262 103852-126 77014-088 77014-090 77014-092 77091-620 77091-622 77304-672
Methyl-5-bromo-2-hydroxynicotinate 98%
Methyl-5-bromo-2-hydroxynicotinate

Precaución: for Research Use Only

Formula: C₇H₆BrNO₃
MW: 232.03 g/mol
Boiling Pt: 346.4 °C (760 mm Hg)
Melting Pt: 181…183 °C
Flash Pt: 163.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03844843
CAS Number: 120034-05-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR