Methyl 5-bromo-2-naphthoate 98%

Supplier: Ambeed
Warning

A100710-100MG A100710-250MG A100710-5G
103795-586EA 283 USD
103795-586 103821-284 76717-502 76717-504 77282-546
Methyl 5-bromo-2-naphthoate 98%
Methyl 5-bromo-2-naphthoate

Precaución: for Research Use Only

Formula: C₁₂H₉BrO₂
MW: 265.1 g/mol
Boiling Pt: 357.0±15.0 °C (760 mm Hg)
Melting Pt: 73 °C
Flash Pt: 169.7 °C
Storage Temperature: Ambient
MDL Number: MFCD08275021
CAS Number: 67878-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR