Methyl-5-bromo-2-methylbenzoate 98%

Supplier: Ambeed
Warning

A121706-100G A121706-10G A121706-500G A121706-5G A121706-25G
76718-576EA 59 USD
76718-576 76718-578 76718-580 77312-246 77566-256
Methyl-5-bromo-2-methylbenzoate 98%
Methyl-5-bromo-2-methylbenzoate

Precaución: For research use only.

Formula: C₉H₉BrO₂
Boiling Pt: 258.9 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD09839555
CAS Number: 79669-50-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR