Methyl-5-bromo-2-iodobenzoate 96%

Supplier: Ambeed
Warning

Synonyms: Methyl 5-bromo-2-iodobenzoate

A239019-100G A239019-10G A239019-5G A239019-1G A239019-25G
103811-258EA 11 USD
103811-258 103851-054 77031-246 77031-248 77090-190 77304-128 77304-130
Methyl-5-bromo-2-iodobenzoate 96%
Methyl 5-bromo-2-iodobenzoate

Precaución: for Research Use Only

Formula: C₈H₆BrIO₂
MW: 340.9400 g/mol
Boiling Pt: 307.1 °C (760 mm Hg)
Melting Pt: 45…49 °C
Flash Pt: 139.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00144771
CAS Number: 181765-86-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR