Methyl-5-bromosalicylate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 5-bromo-2-hydroxybenzoate
A171289-100G
A171289-10G
A171289-500G
A171289-25G
A171289-5G
A171289-1G
103806-618EA
6
USD
103806-618
103844-892
76720-528
76720-530
76720-532
77082-034
77082-036
77301-114
Methyl-5-bromosalicylate 97%
Methyl 5-bromosalicylate
Precaución: for Research Use Only
Formula:
C₈H₇BrO₃ MW: 231.05 g/mol Boiling Pt: 266.6 °C (760 mm Hg) Melting Pt: 61…65 °C Flash Pt: 115 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016460 CAS Number: 4068-76-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...