Methyl-5-bromo-2-fluorobenzoate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 5-bromo-2-fluorobenzoate
A223807-100G
A223807-10G
A223807-1G
A223807-5G
103819-068EA
9
USD
103819-068
103849-950
76826-246
76826-248
77088-576
77088-578
Methyl-5-bromo-2-fluorobenzoate 98%
Methyl 5-bromo-2-fluorobenzoate
Precaución: for Research Use Only
Formula:
C₈H₆BrFO₂ MW: 233.0300 g/mol Flash Pt: 73 °C Storage Temperature: Ambient |
MDL Number:
MFCD06203710 CAS Number: 57381-59-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...