Methyl 5-bromo-2-(trifluoromethyl)isonicotinate 97%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-2-trifluoromethyl-isonicotinic acid methyl ester

A184849-100MG A184849-250MG A184849-1G
103815-796EA 68 USD
103815-796 103846-362 76825-054 76825-056 77083-988
Methyl 5-bromo-2-(trifluoromethyl)isonicotinate 97%
Methyl 5-bromo-2-(trifluoromethyl)isonicotinate

Precaución: for Research Use Only

Formula: C₈H₅BrF₃NO₂
MW: 284.03 g/mol
Boiling Pt: 261.5±40.0 °C (760 mm Hg)
Flash Pt: 112 °C
Storage Temperature: Ambient
MDL Number: MFCD16657895
CAS Number: 1211589-41-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR