Methyl-5-bromo-2-(methylthio)pyrimidine-4-carboxylate 97%

Supplier: Ambeed
Warning

A126554-100G A126554-10G A126554-25G A126554-5G A126554-1G
103800-832EA 5 USD
103800-832 76718-748 76718-750 77298-134 77298-136 77313-054
Methyl-5-bromo-2-(methylthio)pyrimidine-4-carboxylate 97%
Methyl-5-bromo-2-(methylthio)pyrimidine-4-carboxylate

Precaución: for Research Use Only

Formula: C₇H₇BrN₂O₂S
Melting Pt: 67 °C
Storage Temperature: Ambient
MDL Number: MFCD05664428
CAS Number: 50593-91-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR