Methyl 5-bromo-1H-pyrrole-3-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-4-(methoxycarbonyl)-1H-pyrrole

A138576-100MG A138576-250MG A138576-1G
103802-632EA 51 USD
103802-632 76996-792 76996-794 77315-672
Methyl 5-bromo-1H-pyrrole-3-carboxylate 97%
Methyl 5-bromo-1H-pyrrole-3-carboxylate

Precaución: for Research Use Only

Formula: C₆H₆BrNO₂
Flash Pt: 160.148 °C
Storage Temperature: Freezer
MDL Number: MFCD09841652
CAS Number: 16420-39-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR