Methyl-5-bromo-1H-pyrrole-2-carboxylate 95%
Supplier: Ambeed
|
Warning
|
A105551-100MG
A105551-10G
A105551-250MG
A105551-1G
A105551-5G
A105551-25G
103796-898EA
39
USD
103796-898
103822-632
77025-384
77025-386
77025-388
77309-502
77309-504
77871-405
Methyl-5-bromo-1H-pyrrole-2-carboxylate 95%
Methyl-5-bromo-1H-pyrrole-2-carboxylate
Precaución: for Research Use Only
Formula:
C₆H₆BrNO₂ Flash Pt: 129.481 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09801030 CAS Number: 934-07-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...