Methyl 5-bromo-1H-pyrazole-3-carboxylate 97%

Supplier: Ambeed
Warning

A153013-100MG A153013-10G A153013-250MG A153013-1G A153013-5G
103804-458EA 17 USD
103804-458 103842-630 76871-706 76871-708 76871-710 77318-268 77318-270
Methyl 5-bromo-1H-pyrazole-3-carboxylate 97%
Methyl 5-bromo-1H-pyrazole-3-carboxylate

Precaución: for Research Use Only

Formula: C₅H₅BrN₂O₂
MW: 205 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD23701497
CAS Number: 1328893-17-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR