Methyl-5-bromo-1H-indole-3-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 5-bromo-1H-indole-3-carboxylate

A398152-100G A398152-10G A398152-250MG
103827-014EA 10 USD
103827-014 103838-054 77016-276 77684-133 77684-134
Methyl-5-bromo-1H-indole-3-carboxylate 98%
Methyl-5-bromo-1H-indole-3-carboxylate

Precaución: For research use only.

Formula: C₁₀H₈BrNO₂
MW: 254.0800 g/mol
Boiling Pt: 386.2 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD06203728
CAS Number: 773873-77-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR