Methyl 5-bromo-1H-indazole-7-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-7-(methoxycarbonyl)-1H-indazole

A130469-100MG A130469-250MG A130469-5G A130469-1G
103801-428EA 42 USD
103801-428 76972-040 76972-042 77298-408 77313-862
Methyl 5-bromo-1H-indazole-7-carboxylate 98%
Methyl 5-bromo-1H-indazole-7-carboxylate

Precaución: for Research Use Only

Formula: C₉H₇BrN₂O₂
MW: 255.068 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08458991
CAS Number: 898747-24-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR