Methyl-5-bromo-1H-1,2,4-triazole-3-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 5-bromo-1H-1,2,4-triazole-3-carboxylate

A126924-100MG A126924-10G A126924-250MG A126924-1G A126924-25G A126924-5G
103800-874EA 14 USD
103800-874 76971-898 76971-900 76971-902 77313-112 77313-114 77313-116
Methyl-5-bromo-1H-1,2,4-triazole-3-carboxylate 97%
Methyl-5-bromo-1H-1,2,4-triazole-3-carboxylate

Precaución: for Research Use Only

Formula: C₄H₄BrN₃O₂
MW: 206.0000 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13188574
CAS Number: 704911-47-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR