Methyl-5-benzyloxy-4-methoxy-2-nitrobenzoate 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 5-benzyloxy-4-methoxy-2-nitrobenzoate
A173849-10G
A173849-250MG
A173849-25G
A173849-1G
A173849-5G
103806-876EA
35
USD
103806-876
103845-174
76974-158
76974-160
76974-162
77082-460
77082-462
Methyl-5-benzyloxy-4-methoxy-2-nitrobenzoate 95%
Methyl-5-benzyloxy-4-methoxy-2-nitrobenzoate
Precaución: for Research Use Only
Formula:
C₁₆H₁₅NO₆ MW: 317.2900 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD17168894 CAS Number: 164161-49-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...