Methyl-5-aminopicolinate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 5-aminopyridine-2-carboxylate, Methyl 5-aminopicolinate
A360903-1G
A360903-5G
A360903-10G
A360903-250MG
103826-274EA
17
USD
103826-274
103837-286
76840-502
76840-504
77278-312
77278-314
Methyl-5-aminopicolinate 98%
Methyl 5-aminopicolinate
Precaución: for Research Use Only
Formula:
C₇H₈N₂O₂ Boiling Pt: 333.6 °C (760 mm Hg) Flash Pt: 155.587 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD07783939 CAS Number: 67515-76-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...