Methyl 5-aminopentanoate hydrochloride 97%

Supplier: Ambeed
Warning

A633490-25G A633490-250MG A633490-10G
103840-292EA 51 USD
103840-292 76844-470 76980-688 77882-467
Methyl 5-aminopentanoate hydrochloride 97%
Methyl 5-aminopentanoate hydrochloride

Precaución: For research use only

Formula: C₆H₁₃NO₂·ClH
Melting Pt: 145.5…147 °C
Flash Pt: 82.1 °C
Storage Temperature: Freezer
MDL Number: MFCD13368250
CAS Number: 29840-56-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR