Methyl-5-aminonicotinate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 5-aminonicotinate, Methyl 5-aminopyridine-3-carboxylate

A102732-100G A102732-10G A102732-250MG A102732-25G A102732-1G A102732-5G
103796-422EA 15 USD
103796-422 103822-118 77160-298 77160-300 77160-302 77160-304 77308-986 77308-988
Methyl-5-aminonicotinate 97%
Methyl-5-aminonicotinate

Precaución: for Research Use Only

Formula: C₇H₈N₂O₂
MW: 152.1500 g/mol
Melting Pt: 135…137 °C
Flash Pt: 147 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04038688
CAS Number: 36052-25-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR