Methyl 5-amino-2-nitrobenzoate 97%

Supplier: Ambeed
Warning

Synonyms: 5-Amino-2-nitro-benzoic acid methyl ester, Methyl 5-amino-2-nitrobenzoate

A435238-1G A435238-25G
103827-592EA 9 USD
103827-592 103838-660 76842-380 76842-382
Methyl 5-amino-2-nitrobenzoate 97%
Methyl 5-amino-2-nitrobenzoate

Precaución: for Research Use Only

Formula: C₈H₈N₂O₄
MW: 196.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12913482
CAS Number: 35998-96-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR