Methyl-5-aminosalicylate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 5-amino-2-hydroxybenzoate
A285343-100G
A285343-250MG
A285343-1G
A285343-25G
A285343-5G
103813-668EA
11
USD
103813-668
103834-802
76723-390
76723-392
76837-332
76837-334
76837-336
Methyl-5-aminosalicylate 98%
Methyl 5-aminosalicylate
Precaución: for Research Use Only
Formula:
C₈H₉NO₃ MW: 167.1620 g/mol Boiling Pt: 319.2 °C (760 mm Hg) Flash Pt: 146.8 °C Storage Temperature: Ambient |
MDL Number:
MFCD01317557 CAS Number: 42753-75-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...