Methyl 5-amino-2-chloroisonicotinate 97%

Supplier: Ambeed
Warning

Synonyms: 5-Amino-2-chloro-4-(methoxycarbonyl)pyridine, Methyl 5-amino-2-chloropyridine-4-carboxylate

A307870-10G A307870-250MG A307870-1G A307870-25G
103814-526EA 39 USD
103814-526 103835-702 76988-900 76988-902 77305-968 77305-970
Methyl 5-amino-2-chloroisonicotinate 97%
Methyl 5-amino-2-chloroisonicotinate

Precaución: for Research Use Only

Formula: C₇H₇ClN₂O₂
MW: 186.596 g/mol
Boiling Pt: 340.3±37.0 °C (760 mm Hg)
Flash Pt: 159.6 °C
Storage Temperature: Ambient
MDL Number: MFCD11848213
CAS Number: 1073182-59-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR