Methyl-5-amino-1H-pyrazole-3-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 5-amino-1H-pyrazole-3-carboxylate

A146155-100MG A146155-250MG A146155-1G A146155-25G A146155-5G A146155-10G
103803-636EA 26 USD
103803-636 103841-722 77235-206 77235-208 77317-032 77317-034 77317-036 77746-025
Methyl-5-amino-1H-pyrazole-3-carboxylate 95%
Methyl-5-amino-1H-pyrazole-3-carboxylate

Precaución: for Research Use Only

Formula: C₅H₇N₃O₂
MW: 141.1300 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08063165
CAS Number: 632365-54-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR