Methyl-5-acetylsalicylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 5-acetyl-2-hydroxybenzoate

A155325-100G A155325-10G
103842-936EA 6 USD
103842-936 76689-534 76689-536
Methyl-5-acetylsalicylate 98%
Methyl 5-acetylsalicylate

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₄
Boiling Pt: 345.4 °C (760 mm Hg)
Melting Pt: 62…64 °C
Storage Temperature: Ambient
MDL Number: MFCD00040924
CAS Number: 16475-90-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR