Methyl 5,6-dichloropicolinate 97%

Supplier: Ambeed
Warning

A162989-10G A162989-250MG A162989-1G A162989-25G A162989-5G
103805-676EA 19 USD
103805-676 103843-934 76872-098 76872-100 77320-010 77320-012 77320-014
Methyl 5,6-dichloropicolinate 97%
Methyl 5,6-dichloropicolinate

Precaución: for Research Use Only

Formula: C₇H₅Cl₂NO₂
MW: 206.02 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD14698032
CAS Number: 1214375-24-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR