Methyl-5,6-dichloronicotinate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 5,6-dichloronicotinate

A155778-100G A155778-10G A155778-250MG A155778-1G A155778-25G
103804-802EA 6 USD
103804-802 103843-004 76719-940 76719-942 76719-944 77318-742 77318-744
Methyl-5,6-dichloronicotinate 97%
Methyl-5,6-dichloronicotinate

Precaución: for Research Use Only

Formula: C₇H₅Cl₂NO₂
MW: 206.03 g/mol
Boiling Pt: 265.5 °C (760 mm Hg)
Melting Pt: 63…65 °C
Flash Pt: 114.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00125367
CAS Number: 56055-54-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR