Methyl 5-(hydroxymethyl)nicotinate 97%

Supplier: Ambeed
Warning

A185701-250MG A185701-1G A185701-5G
103815-876EA 67 USD
103815-876 103846-446 76769-728 77084-078 77084-080
Methyl 5-(hydroxymethyl)nicotinate 97%
Methyl 5-(hydroxymethyl)nicotinate

Precaución: for Research Use Only

Formula: C₈H₉NO₃
MW: 167.16 g/mol
Boiling Pt: 316.3±32.0 °C (760 mm Hg)
Flash Pt: 145.1 °C
Storage Temperature: Ambient
MDL Number: MFCD11848097
CAS Number: 129747-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR