Methyl 5-(bromomethyl)picolinate 96%

Supplier: Ambeed
Danger

A324351-100MG A324351-250MG A324351-50MG
103825-368EA 458 USD
103825-368 103836-312 77668-446 77879-673 77879-674
Methyl 5-(bromomethyl)picolinate 96%
Methyl 5-(bromomethyl)picolinate

Precaución: Inert atmosphere, 2…8 °C.

Formula: C₈H₈BrNO₂
MW: 230.05 g/mol
Boiling Pt: 334 °C (760 mm Hg)
Flash Pt: 156 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07367952
CAS Number: 55876-84-1
UN: 1759
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR