Methyl-4-oxotetrahydrofuran-3-carboxylate 96%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-oxotetrahydrofuran-3-carboxylate

A177266-100MG A177266-250MG A177266-1G A177266-5G A177266-10G
103807-220EA 35 USD
103807-220 103845-548 76974-294 76974-296 77082-938 77082-940 77550-138
Methyl-4-oxotetrahydrofuran-3-carboxylate 96%
Methyl-4-oxotetrahydrofuran-3-carboxylate

Precaución: for Research Use Only

Formula: C₆H₈O₄
MW: 144.13 g/mol
Boiling Pt: 220 °C (760 mm Hg)
Flash Pt: 91 °C
Storage Temperature: Refrigerator
MDL Number: MFCD19707049
CAS Number: 57595-23-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR