Methyl tetrahydro-4-oxo-2H-thiopyran-3-carboxylate 97%, Ambeed.Inc

Supplier: Ambeed
Warning

A120166-250MG A120166-1G A120166-5G A120166-10G
103799-896EA 32 USD
103799-896 77009-406 77312-000 77312-002 77683-079
Methyl tetrahydro-4-oxo-2H-thiopyran-3-carboxylate 97%, Ambeed.Inc
Methyl tetrahydro-4-oxo-2H-thiopyran-3-carboxylate

Precaución: For research use only.

Formula: C₇H₁₀O₃S
Boiling Pt: 285 °C (760 mm Hg)
Melting Pt: 62…66 °C
Flash Pt: 137.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00203485
CAS Number: 4160-61-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR