Methyl-4-oxotetrahydro-2H-pyran-3-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-oxotetrahydro-2H-pyran-3-carboxylate

A106668-100MG A106668-1G A106668-5G A106668-10G A106668-250MG
103797-056EA 52 USD
103797-056 103822-812 76809-670 77309-668 77309-670 77570-398 77670-528
Methyl-4-oxotetrahydro-2H-pyran-3-carboxylate 95%
Methyl-4-oxotetrahydro-2H-pyran-3-carboxylate

Precaución: for Research Use Only

Formula: C₇H₁₀O₄
MW: 158.16 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20921984
CAS Number: 127956-11-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR