Methyl-4-oxopiperidine-3-carboxylate hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-oxopiperidine-3-carboxylate hydrochloride, Methyl 4-Oxopiperidine-3-carboxylate HCl

A485065-100G A485065-10G A485065-1G A485065-25G
103828-168EA 5 USD
103828-168 103839-242 76725-924 76725-926 76843-180 76843-182
Methyl-4-oxopiperidine-3-carboxylate hydrochloride 98%
Methyl-4-oxopiperidine-3-carboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₂ClNO₃
MW: 193.63 g/mol
Melting Pt: 175 °C
Storage Temperature: Ambient
MDL Number: MFCD00012769
CAS Number: 71486-53-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR