Methyl-4-methylpicolinate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 4-methylpicolinate
A240169-250MG
A240169-5G
103811-332EA
14
USD
103811-332
103851-130
76976-436
77090-280
Methyl-4-methylpicolinate 97%
Methyl 4-methylpicolinate
Precaución: for Research Use Only
Formula:
C₈H₉NO₂ MW: 151.1600 g/mol Flash Pt: 111.487 °C Storage Temperature: Ambient |
MDL Number:
MFCD09265118 CAS Number: 13509-13-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...