Methyl-4-methylnicotinate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-methylnicotinate

A174666-1G A174666-5G A174666-25G
103806-966EA 30 USD
103806-966 103845-266 77082-572 77082-574 77301-350
Methyl-4-methylnicotinate 98%
Methyl-4-methylnicotinate

Precaución: for Research Use Only

Formula: C₈H₉NO₂
MW: 151.1600 g/mol
Boiling Pt: 215.2 °C (760 mm Hg)
Flash Pt: 84 °C
Storage Temperature: Ambient
MDL Number: MFCD09258869
CAS Number: 33402-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR