Methyl-4-methoxynicotinate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-Methoxynicotinate

A667525-100MG A667525-10G A667525-250MG A667525-5G A667525-1G
103840-482EA 174 USD
103840-482 76727-482 76727-484 76727-486 76844-686 77308-080
Methyl-4-methoxynicotinate 98%
Methyl-4-methoxynicotinate

Precaución: for Research Use Only

Formula: C₈H₉NO₃
MW: 167.16 g/mol
Boiling Pt: 250.9 °C (760 mm Hg)
Flash Pt: 105.5 °C
Storage Temperature: Ambient
MDL Number: MFCD09864940
CAS Number: 10177-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR