Methyl-4-methoxy-2-methylbenzoate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-methoxy-2-methylbenzoate

A344306-1G A344306-25G A344306-5G A344306-10G
103825-902EA 30 USD
103825-902 103836-882 76839-930 76839-932 76839-934 76978-446
Methyl-4-methoxy-2-methylbenzoate 97%
Methyl-4-methoxy-2-methylbenzoate

Precaución: For research use only

Formula: C₁₀H₁₂O₃
MW: 180.21 g/mol
Boiling Pt: 265.0±20.0 °C (760 mm Hg)
Flash Pt: 105.4 °C
Storage Temperature: Ambient
MDL Number: MFCD06203918
CAS Number: 35598-05-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR